Learn More
Medchemexpress LLC N-Boc-biocytin | 62062-43-5 | MFCD00133628 | >97.0% | 472.60 | C21H36N4O6S | 25 MG
SDP
Catalog No. 5000282525
Encompass_Preferred
N-Boc-biocytin is a biotin-PEG linker bearing a carboxylic acid and a Boc-protected amine. It is used to attach biotin to primary amines via standard amide-coupling chemistry while protecting the linker's amine during multi-step syntheses. The compound is supplied as a white to off-white solid and is suitable for affinity tagging, probe synthesis, and general bioconjugation workflows.
  • Protects amine functionality during multi-step synthesis.
  • Carboxylic acid enables amide coupling to primary amines.
  • Provides biotin for affinity capture and detection.
  • Available in multiple small-scale sizes for research use.
  • Solid form simplifies handling and accurate weighing.
  • Stable when stored under recommended conditions.
Catalog No. 50-002-82525 Supplier Medchemexpress LLC Supplier No. HYW28661325MG
N-Boc-biocytin is a biotin-PEG linker bearing a carboxylic acid and a Boc-protected amine. It is used to attach biotin to primary amines via standard amide-coupling chemistry while protecting the linker's amine during multi-step syntheses. The compound is supplied as a white to off-white solid and is suitable for affinity tagging, probe synthesis, and general bioconjugation workflows.
  • Protects amine functionality during multi-step synthesis.
  • Carboxylic acid enables amide coupling to primary amines.
  • Provides biotin for affinity capture and detection.
  • Available in multiple small-scale sizes for research use.
  • Solid form simplifies handling and accurate weighing.
  • Stable when stored under recommended conditions.