Learn More
Medchemexpress LLC N-Boc-biocytin | 62062-43-5 | MFCD00133628 | >97.0% | 472.60 g/mol | C21H36N4O6S | 50 MG
SDP
Catalog No. 5000280597
Encompass_Preferred
N-Boc-Biocytin is a Boc-protected biotin PEG linker bearing a terminal carboxylic acid; it enables conjugation to primary amines through stable amide bond formation. The Boc group protects the amine during synthesis and can be cleaved under acidic conditions to reveal a free amine for subsequent derivatization. This reagent is intended for research use in peptide synthesis and bioconjugation workflows where a protected biotin handle is required.
  • Boc-protected amine for controlled deprotection
  • Terminal carboxylic acid for stable amide bond formation with primary amines
  • Biotin functionality for affinity tagging and detection
  • PEG spacer improves solubility and reduces steric hindrance
  • Suitable for peptide synthesis and bioconjugation applications
Catalog No. 50-002-80597 Supplier Medchemexpress LLC Supplier No. HYW28661350MG
N-Boc-Biocytin is a Boc-protected biotin PEG linker bearing a terminal carboxylic acid; it enables conjugation to primary amines through stable amide bond formation. The Boc group protects the amine during synthesis and can be cleaved under acidic conditions to reveal a free amine for subsequent derivatization. This reagent is intended for research use in peptide synthesis and bioconjugation workflows where a protected biotin handle is required.
  • Boc-protected amine for controlled deprotection
  • Terminal carboxylic acid for stable amide bond formation with primary amines
  • Biotin functionality for affinity tagging and detection
  • PEG spacer improves solubility and reduces steric hindrance
  • Suitable for peptide synthesis and bioconjugation applications