Learn More
Medchemexpress LLC N-Boc-biocytin | 62062-43-5 | MFCD00133628 | ≥97.0% | 472.60 g/mol | C21H36N4O6S | 500 MG
SDP
Catalog No. 5000280596
Encompass_Preferred
N-Boc-Biocytin is a Boc-protected biotin PEG linker containing a carboxylic acid and a Boc-protected primary amine. The carboxylic group reacts with primary amines to form stable, irreversible amide bonds, and the Boc group can be deprotected under acidic conditions to reveal a free amine for further conjugation. Supplied as a white to off-white solid with storage and shipping guidance for reagent stability.
  • Provides a biotinylated PEG linker for conjugation chemistry.
  • Forms stable amide bonds with primary amines.
  • Boc protecting group allows controlled deprotection under acidic conditions.
  • White to off-white solid suitable for biochemical assays.
  • Stable when stored at recommended temperatures for extended shelf life.
Catalog No. 50-002-80596 Supplier Medchemexpress LLC Supplier No. HYW286613500MG
N-Boc-Biocytin is a Boc-protected biotin PEG linker containing a carboxylic acid and a Boc-protected primary amine. The carboxylic group reacts with primary amines to form stable, irreversible amide bonds, and the Boc group can be deprotected under acidic conditions to reveal a free amine for further conjugation. Supplied as a white to off-white solid with storage and shipping guidance for reagent stability.
  • Provides a biotinylated PEG linker for conjugation chemistry.
  • Forms stable amide bonds with primary amines.
  • Boc protecting group allows controlled deprotection under acidic conditions.
  • White to off-white solid suitable for biochemical assays.
  • Stable when stored at recommended temperatures for extended shelf life.