Learn More
Medchemexpress LLC N-Boc-biocytin | 62062-43-5 | MFCD00133628 | >98.0% | 472.60 g·mol⁻¹ | C21H36N4O6S | 100 MG
SDP
Catalog No. 5000282019
Encompass_Preferred
N-Boc-Biocytin is a Boc-protected biotin-PEG linker containing a carboxylic acid and a Boc-protected primary amine. It permits formation of stable amide bonds with primary amines and, after acidic removal of the Boc group, provides a free amine for further conjugation. The reagent is intended for biotinylation and affinity tagging of biomolecules.
Catalog No. 50-002-82019 Supplier Medchemexpress LLC Supplier No. HYW286613100MG
N-Boc-Biocytin is a Boc-protected biotin-PEG linker containing a carboxylic acid and a Boc-protected primary amine. It permits formation of stable amide bonds with primary amines and, after acidic removal of the Boc group, provides a free amine for further conjugation. The reagent is intended for biotinylation and affinity tagging of biomolecules.