Learn More
Medchemexpress LLC N-boc-biocytin | 62062-43-5 | MFCD00133628 | >=98.0% | 472.60 g/mol | C21H36N4O6S | 250 MG
SDP
Catalog No. 5000282524
Encompass_Preferred
N-boc-biocytin is a Boc-protected biotin-PEG linker containing a carboxylic acid and a Boc-protected primary amine. It is designed for coupling to primary amines to form stable amide bonds; the Boc protecting group can be removed under acidic conditions to reveal a free amine for further conjugation and biotinylation applications.
  • Biotin-PEG linker with carboxylic acid and Boc-protected primary amine.
  • Enables formation of stable, irreversible amide bonds with primary amines.
  • Boc group removable under acidic conditions to expose a reactive amine.
  • Suitable for biotinylation and conjugation in protein and peptide chemistry.
  • Molecular weight 472.60 g/mol; formula C21H36N4O6S; CAS 62062-43-5.
  • Available in multiple pack sizes to support research workflows.
Catalog No. 50-002-82524 Supplier Medchemexpress LLC Supplier No. HYW286613250MG
N-boc-biocytin is a Boc-protected biotin-PEG linker containing a carboxylic acid and a Boc-protected primary amine. It is designed for coupling to primary amines to form stable amide bonds; the Boc protecting group can be removed under acidic conditions to reveal a free amine for further conjugation and biotinylation applications.
  • Biotin-PEG linker with carboxylic acid and Boc-protected primary amine.
  • Enables formation of stable, irreversible amide bonds with primary amines.
  • Boc group removable under acidic conditions to expose a reactive amine.
  • Suitable for biotinylation and conjugation in protein and peptide chemistry.
  • Molecular weight 472.60 g/mol; formula C21H36N4O6S; CAS 62062-43-5.
  • Available in multiple pack sizes to support research workflows.