Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Boc-Cadaverine

Catalog No. 154061ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-406-1ML 1 mL
15-406-5ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 15-406-1ML Supplier Sigma Aldrich Supplier No. 154061ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Some of the reported applications of N-Boc-cadaverine include:Synthesis of of a supermacrocycle that self-assemble to form organic nanotubes.Preparation of water-soluble unsymmetrical sulforhodamine fluorophores from monobrominated sulfoxanthene dye.Synthesis of functionalized porphyrins as biocompatible carrier system for photodynamic therapy (PDT).Building block for preparing polyamines and polyamides

Specifications

CAS 51644-96-3
Density 0.972 g/mL at 20°C
Molecular Formula C10H22N2O2
Linear Formula (CH)3COCONH(CH2)5NH2
Refractive Index n20/D 1.460
Quantity 1 mL
MDL Number MFCD00210020
Synonym N-Boc-1,5-diaminopentane; tert-Butyl N-(5-aminopentyl)carbamate
Molecular Weight (g/mol) 202.29 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.