Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-D-aspartic acid 4-benzyl ester, 98%

Catalog No. AAH6200014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6200014 25 g
AAH6200006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6200014 Supplier Thermo Scientific Chemicals Supplier No. H6200014
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 51186-58-4 | C16H21NO6 | 323.345 g/mol

N-Boc-D-aspartic acid 4-benzyl ester is used as an intermediate in organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-D-aspartic acid 4-benzyl ester is used as an intermediate in organic synthesis and pharmaceuticals.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51186-58-4
Molecular Formula C16H21NO6
Molecular Weight (g/mol) 323.345
MDL Number MFCD00038255
InChI Key SOHLZANWVLCPHK-GFCCVEGCSA-N
Synonym boc-d-asp obzl-oh, boc-d-aspartic acid 4-benzyl ester, r-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, 2r-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-d-aspartic acid beta-benzyl ester, boc-d-asparticacid4-benzylester, pubchem11179, boc-d-asp obn-oh, boc-d-aspartic acid b-benzyl ester, boc-d-aspartic acid ?-benzyl ester
PubChem CID 6994663
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)OCC1=CC=CC=C1)C(=O)O

Specifications

Quantity 25 g
Beilstein 2305471
Formula Weight 323.34
Percent Purity 98%
Chemical Name or Material N-Boc-D-aspartic acid 4-benzyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.