Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Boc-D-glutamic acid 1-tert-butyl ester | 73872-71-6 | MFCD00076926 | >97.0% | 303.35 g/mol | C14H25NO6 | 25 G
SDP

Catalog No. 5000209543
Encompass_Preferred

Boc-D-Glu-OtBu is a Boc- and tert-butyl ester-protected derivative of D-glutamic acid used as a building block in peptide synthesis and other organic syntheses that require temporary protection of the amino and side-chain carboxyl groups.

  • Protected amino acid for peptide synthesis.
  • Features Boc protection on the amino group and tert-butyl ester protection on the side-chain carboxyl.
  • Purity ≥97.0% as supplied.
  • Molecular formula C14H25NO6; molecular weight 303.35 g/mol.
  • Available in 25 G packaging suitable for research-scale synthesis.

Catalog No. 50-002-09543 Supplier Medchemexpress LLC Supplier No. 25GHYW089233
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-D-Glu-OtBu is a Boc- and tert-butyl ester-protected derivative of D-glutamic acid used as a building block in peptide synthesis and other organic syntheses that require temporary protection of the amino and side-chain carboxyl groups.

  • Protected amino acid for peptide synthesis.
  • Features Boc protection on the amino group and tert-butyl ester protection on the side-chain carboxyl.
  • Purity ≥97.0% as supplied.
  • Molecular formula C14H25NO6; molecular weight 303.35 g/mol.
  • Available in 25 G packaging suitable for research-scale synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.