Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-D-glutamic acid 5-benzyl ester, 98%

Catalog No. AAH6253706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6253706 5 g
AAH6253714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6253706 Supplier Thermo Scientific Chemicals Supplier No. H6253706
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 35793-73-8 | C17H23NO6 | 337.372 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35793-73-8
Molecular Formula C17H23NO6
Molecular Weight (g/mol) 337.372
MDL Number MFCD00038258
InChI Key AJDUMMXHVCMISJ-CYBMUJFWSA-N
Synonym boc-d-glu obzl-oh, boc-d-glutamic acid 5-benzyl ester, boc-d-glutamic acid 5-benzylester, n-boc-d-glutamic acid 5-benzyl ester, 2r-5-benzyloxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, ambotzbaa1354, boc-d-glutamic acid gamma-benzyl ester, pubchem12172, pubchem14934, boc-d-glu obn-oh
PubChem CID 7010513
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-phenylmethoxypentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCC(=O)OCC1=CC=CC=C1)C(=O)O

Specifications

Quantity 5 g
Formula Weight 337.37
Percent Purity 98%
Chemical Name or Material N-Boc-D-glutamic acid 5-benzyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.