Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Boc-D-pyroglutamic acid ethyl ester | 144978-35-8 | 257.28 | 100 G
SDP

Catalog No. 5000448695
Encompass_Preferred

Boc-D-Pyr-Oet, also known as N-Boc-D-pyroglutamic acid ethyl ester, is a biochemical assay reagent. It is a solid with a white to off-white appearance.

  • Used as a biochemical assay reagent.
  • Solid, white to off-white appearance.
  • Store at room temperature for 3 years.
  • Store in solvent at -80°C for 2 years or -20°C for 1 year.
  • Soluble in DMSO at 200 mg/mL.

Catalog No. 50-004-48695 Supplier Medchemexpress LLC Supplier No. HYW003605100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-D-Pyr-Oet, also known as N-Boc-D-pyroglutamic acid ethyl ester, is a biochemical assay reagent. It is a solid with a white to off-white appearance.

  • Used as a biochemical assay reagent.
  • Solid, white to off-white appearance.
  • Store at room temperature for 3 years.
  • Store in solvent at -80°C for 2 years or -20°C for 1 year.
  • Soluble in DMSO at 200 mg/mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.