Learn More
Medchemexpress LLC N-Boc-diethanolamine | 103898-11-9 | 100.0% | 205.25 | C9H19NO4 | 100 G
SDP
Catalog No. 5000204116
Encompass_Preferred
N-Boc-diethanolamine is a Boc-protected diethanolamine used as a building block and cleavable linker in organic synthesis and bioconjugation applications. It is supplied for research use at high purity and is suitable for use in linker and small-molecule assembly workflows.
  • Boc-protected secondary amine suitable for selective deprotection
  • Two hydroxyl handles for conjugation or further functionalization
  • Appropriate for use as a cleavable ADC/PROTAC linker component
  • High purity for research and synthetic applications
  • Available in multiple pack sizes, including a 100 G pack
Catalog No. 50-002-04116 Supplier Medchemexpress LLC Supplier No. HYW044078100G
N-Boc-diethanolamine is a Boc-protected diethanolamine used as a building block and cleavable linker in organic synthesis and bioconjugation applications. It is supplied for research use at high purity and is suitable for use in linker and small-molecule assembly workflows.
  • Boc-protected secondary amine suitable for selective deprotection
  • Two hydroxyl handles for conjugation or further functionalization
  • Appropriate for use as a cleavable ADC/PROTAC linker component
  • High purity for research and synthetic applications
  • Available in multiple pack sizes, including a 100 G pack