Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-DL-beta-proline ethyl ester, 97%, Thermo Scientific™

Catalog No. AAH5743003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5743003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5743003 Supplier Thermo Scientific Chemicals Supplier No. H5743003

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170844-49-2
Molecular Formula C12H21NO4
Molecular Weight (g/mol) 243.30
MDL Number MFCD03791256
InChI Key PKDQVUYUWUHNMG-UHFFFAOYNA-N
Synonym ethyl 1-boc-3-pyrrolidinecarboxylate, 1-tert-butyl 3-ethyl pyrrolidine-1,3-dicarboxylate, pyrrolidine-1,3-dicarboxylic acid 1-tert-butyl ester 3-ethyl ester, ethyl 1-boc-beta-prolinate, ethyl 1-boc-3-pyrrolidine-carboxylate, n-boc-dl-beta-proline ethyl ester, 1,3-pyrrolidinedicarboxylic acid, 1-1,1-dimethylethyl 3-ethyl ester, 1-tert-butyl 3-ethylpyrrolidine-1,3-dicarboxylate, pubchem14290, tert-butyl ethyl pyrrolidine-1,3-dicarboxylate
PubChem CID 4192972
IUPAC Name 1-O-tert-butyl 3-O-ethyl pyrrolidine-1,3-dicarboxylate
SMILES CCOC(=O)C1CCN(C1)C(=O)OC(C)(C)C

Specifications

Quantity 1 g
Formula Weight 243.3
Percent Purity 97%
Chemical Name or Material N-Boc-DL-beta-proline ethyl ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.