Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-DL-valine, 98%, Thermo Scientific™

Catalog No. AAH6190814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6190814 25 g
AAH6190806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6190814 Supplier Thermo Scientific Chemicals Supplier No. H6190814

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54895-12-4
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.27
MDL Number MFCD00069973
InChI Key SZXBQTSZISFIAO-UHFFFAOYSA-N
Synonym boc-dl-valine, boc-dl-val-oh, n-boc-dl-valine, 2-tert-butoxycarbonyl amino-3-methylbutanoic acid, dl-n-tert-butoxycarbonyl valine, n-tert-butoxycarbonyl valine, 2-tert-butoxy carbonyl amino-3-methylbutanoic acid, n-tert-butoxycarbonyl-dl-valine, n-alpha-t-boc-l-valine
PubChem CID 270655
IUPAC Name 3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(C)C(NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 112°C to 114°C
Beilstein 9196510
Quantity 25 g
Formula Weight 217.26
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Boc-DL-valine
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.