Learn More
Medchemexpress LLC N-Boc-dolaproine | 120205-50-7 | 99.96% | 287.36 | 500 MG
SDP
Catalog No. 5000401446
Encompass_Preferred
N-Boc-dolaproine ((2R,3R)-BOC-dolaproine) is the amino acid residue of the pentapeptide Dolastatin 10. Dolastatin 10 inhibits tubulin polymerization and mitosis and has anticancer activity. N-Boc-dolaproine is a key intermediate used in the synthesis of Dolastatin 10.
  • Amino acid residue of Dolastatin 10
  • Inhibits tubulin polymerization
  • Inhibits mitosis
  • Has anticancer activity
  • Key intermediate for Dolastatin 10 synthesis
Catalog No. 50-004-01446 Supplier Medchemexpress LLC Supplier No. HY33046500MG
N-Boc-dolaproine ((2R,3R)-BOC-dolaproine) is the amino acid residue of the pentapeptide Dolastatin 10. Dolastatin 10 inhibits tubulin polymerization and mitosis and has anticancer activity. N-Boc-dolaproine is a key intermediate used in the synthesis of Dolastatin 10.
  • Amino acid residue of Dolastatin 10
  • Inhibits tubulin polymerization
  • Inhibits mitosis
  • Has anticancer activity
  • Key intermediate for Dolastatin 10 synthesis