Learn More
Medchemexpress LLC N-Boc-dolaproine-amide-Me-Phe | 3026596-72-2 | 98.5% | 404.54 g/mol | C23H36N2O4 | 10 MG
SDP
Catalog No. 5000298330
Encompass_Preferred
N-Boc-dolaproine-amide-Me-Phe is a Boc-protected amino acid fragment derived from the pentapeptide Dolastatin 10, provided as a high-purity research reagent for studies of tubulin polymerization and antimitotic activity.
  • High purity suitable for research applications.
  • Available as a solid and as a 10 mM solution in DMSO.
  • Stable when stored sealed at 4°C; in solution, store at -80°C for long-term storage.
  • Light yellow solid appearance.
  • Commonly used for peptide synthesis and structure-activity studies.
Catalog No. 50-002-98330 Supplier Medchemexpress LLC Supplier No. HY4905310MG
N-Boc-dolaproine-amide-Me-Phe is a Boc-protected amino acid fragment derived from the pentapeptide Dolastatin 10, provided as a high-purity research reagent for studies of tubulin polymerization and antimitotic activity.
  • High purity suitable for research applications.
  • Available as a solid and as a 10 mM solution in DMSO.
  • Stable when stored sealed at 4°C; in solution, store at -80°C for long-term storage.
  • Light yellow solid appearance.
  • Commonly used for peptide synthesis and structure-activity studies.