Learn More
Medchemexpress LLC N-Boc-dolaproine-amide-Me-Phe | 3026596-72-2 | 98.5% | C23H36N2O4 | 100 MG
SDP
Catalog No. 5000357838
Encompass_Preferred
N-Boc-dolaproine-amide-Me-Phe is an amino acid residue of the pentapeptide Dolastatin 10, which is known for its ability to inhibit tubulin polymerization and mitosis, demonstrating anticancer activity.
  • Amino acid residue of the pentapeptide Dolastatin 10.
  • Dolastatin 10 inhibits tubulin polymerization and mitosis.
  • Exhibits anticancer activity.
  • Intended for research use only.
Catalog No. 50-003-57838 Supplier Medchemexpress LLC Supplier No. HY49053100MG
N-Boc-dolaproine-amide-Me-Phe is an amino acid residue of the pentapeptide Dolastatin 10, which is known for its ability to inhibit tubulin polymerization and mitosis, demonstrating anticancer activity.
  • Amino acid residue of the pentapeptide Dolastatin 10.
  • Dolastatin 10 inhibits tubulin polymerization and mitosis.
  • Exhibits anticancer activity.
  • Intended for research use only.