Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Boc-dolaproine-amide-Me-Phe | 3026596-72-2 | 98.5% | C23H36N2O4 | 50 MG
SDP

Catalog No. 5000359269
Encompass_Preferred

N-Boc-dolaproine-amide-Me-Phe is an amino acid residue of the pentapeptide Dolastatin 10. Dolastatin 10 inhibits tubulin polymerization and mitosis, demonstrating anticancer activity. This product is suitable for research use.

  • Amino acid residue of pentapeptide Dolastatin 10
  • Inhibits tubulin polymerization
  • Inhibits mitosis
  • Possesses anticancer activity
  • Molecular weight: 404.54
  • Appearance: Light yellow to yellow solid
  • Storage: 4°C sealed, away from moisture
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Solubility in DMSO: 200 mg/mL

Catalog No. 50-003-59269 Supplier Medchemexpress LLC Supplier No. HY4905350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Boc-dolaproine-amide-Me-Phe is an amino acid residue of the pentapeptide Dolastatin 10. Dolastatin 10 inhibits tubulin polymerization and mitosis, demonstrating anticancer activity. This product is suitable for research use.

  • Amino acid residue of pentapeptide Dolastatin 10
  • Inhibits tubulin polymerization
  • Inhibits mitosis
  • Possesses anticancer activity
  • Molecular weight: 404.54
  • Appearance: Light yellow to yellow solid
  • Storage: 4°C sealed, away from moisture
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Solubility in DMSO: 200 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.