Learn More
Medchemexpress LLC N-Boc-dolaproine dicyclohexylamine | 1369427-40-6 | 99.9% | 468.67 | 500 MG
SDP
Catalog No. 5000400422
Encompass_Preferred
N-Boc-dolaproine dicyclohexylamine is an amino acid residue of the pentapeptide Dolastatin 10. Dolastatin 10 inhibits tubulin polymerization and mitosis and has anticancer activity. This product also contains dicyclohexylamine and is intended for research use only.
  • Inhibits tubulin polymerization
  • Inhibits mitosis
  • Possesses anticancer activity
  • Suitable for laboratory research applications
Catalog No. 50-004-00422 Supplier Medchemexpress LLC Supplier No. HY33046A500MG
N-Boc-dolaproine dicyclohexylamine is an amino acid residue of the pentapeptide Dolastatin 10. Dolastatin 10 inhibits tubulin polymerization and mitosis and has anticancer activity. This product also contains dicyclohexylamine and is intended for research use only.
  • Inhibits tubulin polymerization
  • Inhibits mitosis
  • Possesses anticancer activity
  • Suitable for laboratory research applications