Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Boc-Ethanolamine

Catalog No. 38202725ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-202-725ML 25 mL
38-202-75ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 38-202-725ML Supplier Sigma Aldrich Supplier No. 38202725ML
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Amine protected, difunctional reagent employed in the synthesis of phosphatidyl ethanolamines and ornithine.

Specifications

CAS 26690-80-2
Density 1.042 g/mL at 25°C
Molecular Formula C7H15NO3
Linear Formula (CH3)3COCONHCH2CH2OH
Refractive Index n20/D 1.449
MDL Number MFCD00056657
Quantity 25 mL
Synonym N-(tert-Butoxycarbonyl)ethanolamine; tert-Butyl N-(2-hydroxyethyl)carbamate; Boc-2-aminoethanol; Boc-glycinol
Molecular Weight (g/mol) 161.2 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.