Learn More
Medchemexpress LLC N-Boc-indole-2-boronic acid | 213318-44-6 | MFCD02093045 | 99.95% | C13H16BNO4 | 10 G
SDP
Catalog No. 5000357943
Encompass_Preferred
(1-(tert-Butoxycarbonyl)-1H-indol-2-yl)boronic acid, also known as N-Boc-indole-2-boronic acid, is a high-purity biochemical reagent with a molecular formula of C13H16BNO4 and a molecular weight of 261.08. It is commonly used as a biological material or organic compound in various life science-related research applications.
  • Biochemical reagent for life science research
  • Organic compound suitable for various applications
  • High purity of 99.95%
  • Stable under recommended storage conditions: -20°C, sealed, away from moisture
Catalog No. 50-003-57943 Supplier Medchemexpress LLC Supplier No. HY5028410G
(1-(tert-Butoxycarbonyl)-1H-indol-2-yl)boronic acid, also known as N-Boc-indole-2-boronic acid, is a high-purity biochemical reagent with a molecular formula of C13H16BNO4 and a molecular weight of 261.08. It is commonly used as a biological material or organic compound in various life science-related research applications.
  • Biochemical reagent for life science research
  • Organic compound suitable for various applications
  • High purity of 99.95%
  • Stable under recommended storage conditions: -20°C, sealed, away from moisture