Learn More
Medchemexpress LLC N-Boc-L-3,5-difluorophenylalanine | 205445-52-9 | MFCD00797555 | 99.8% | 301.29 g·mol-1 | C14H17F2NO4 | 500 MG
SDP
Catalog No. 5000300365
Encompass_Preferred
N-Boc-L-3,5-difluorophenylalanine is a Boc-protected derivative of L-phenylalanine bearing fluorine atoms at the 3 and 5 positions of the aromatic ring. It is supplied as a white to off-white solid and is commonly used as a synthetic building block to introduce difluorophenylalanine residues in peptides for medicinal chemistry and structure-activity relationship studies.
  • Boc-protected amino acid for peptide synthesis.
  • 3,5-difluoro substitution to modulate lipophilicity and metabolic stability.
  • High reported purity suitable for research applications.
  • Solid form for convenient storage and handling.
  • Applicable to medicinal chemistry and SAR studies.
Catalog No. 50-003-00365 Supplier Medchemexpress LLC Supplier No. HYW037441500MG
N-Boc-L-3,5-difluorophenylalanine is a Boc-protected derivative of L-phenylalanine bearing fluorine atoms at the 3 and 5 positions of the aromatic ring. It is supplied as a white to off-white solid and is commonly used as a synthetic building block to introduce difluorophenylalanine residues in peptides for medicinal chemistry and structure-activity relationship studies.
  • Boc-protected amino acid for peptide synthesis.
  • 3,5-difluoro substitution to modulate lipophilicity and metabolic stability.
  • High reported purity suitable for research applications.
  • Solid form for convenient storage and handling.
  • Applicable to medicinal chemistry and SAR studies.