Learn More
Medchemexpress LLC N-Boc-L-3,5-difluorophenylalanine | 205445-52-9 | MFCD00797555 | 99.8% | 301.29 g/mol | C14H17F2NO4 | 100 MG
SDP
Catalog No. 5000301159
Encompass_Preferred
N-Boc-L-3,5-difluorophenylalanine is a Boc-protected L-phenylalanine derivative used as a building block in peptide synthesis and medicinal chemistry. Supplied as a white to off-white solid, it has molecular formula C14H17F2NO4 and molecular weight 301.29 g/mol. Recommended storage preserves integrity for extended laboratory use.
  • Boc-protected amino acid for incorporation into peptides.
  • 3,5-difluorophenyl substitution to modulate lipophilicity and metabolic stability.
  • High purity (99.8%) suitable for research applications.
  • Solid powder form for easy handling and weighing.
  • Recommended storage conditions preserve stability (powder: -20°C or 4°C; in solvent: -80°C).
  • Available in small laboratory pack sizes for synthesis workflows.
Catalog No. 50-003-01159 Supplier Medchemexpress LLC Supplier No. HYW037441100MG
N-Boc-L-3,5-difluorophenylalanine is a Boc-protected L-phenylalanine derivative used as a building block in peptide synthesis and medicinal chemistry. Supplied as a white to off-white solid, it has molecular formula C14H17F2NO4 and molecular weight 301.29 g/mol. Recommended storage preserves integrity for extended laboratory use.
  • Boc-protected amino acid for incorporation into peptides.
  • 3,5-difluorophenyl substitution to modulate lipophilicity and metabolic stability.
  • High purity (99.8%) suitable for research applications.
  • Solid powder form for easy handling and weighing.
  • Recommended storage conditions preserve stability (powder: -20°C or 4°C; in solvent: -80°C).
  • Available in small laboratory pack sizes for synthesis workflows.