Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-L-aspartic acid 1-methyl ester, 98%

Catalog No. AAH6259003
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6259003 1 g
AAH6259006 5 g
AAH62590MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6259003 Supplier Thermo Scientific Chemicals Supplier No. H6259003
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 98045-03-5 | C10H17NO6 | 247.247 g/mol

N-Boc-L-aspartic acid 1-methyl ester is used as an intermediate in the preparation of modified alfa-amino acids and 5-cis-alkylprolines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-aspartic acid 1-methyl ester is used as an intermediate in the preparation of modified alfa-amino acids and 5-cis-alkylprolines.

Notes
Incompatible with oxidizing agents. Heat sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98045-03-5
Molecular Formula C10H17NO6
Molecular Weight (g/mol) 247.247
MDL Number MFCD03094777
InChI Key IWFIVTBTZUCTQH-LURJTMIESA-N
Synonym boc-asp-ome, 1-methyl n-boc-l-aspartate, n-boc-l-aspartic acid 1-methyl ester, 3s-3-tert-butoxy carbonyl amino-4-methoxy-4-oxobutanoic acid, n-boc asp oh ome, 1-methyl n-tert-butoxycarbonyl-l-aspartate, n-tert-butoxycarbonylaspartic acid a-methyl ester, n-tert-butoxycarbonyl aspartic acid 1-methyl ester, n-tert-butoxycarbonyl-l-aspartic acid 1-methyl est, n-tert-butoxycarbonyl-l-aspartic acid 1-methyl ester
PubChem CID 11021202
IUPAC Name (3S)-4-methoxy-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)OC

Specifications

Quantity 1 g
Formula Weight 247.25
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Boc-L-aspartic acid 1-methyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.