Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-L-aspartic acid 4-cyclohexyl ester, 95%, Thermo Scientific™

Catalog No. AAH6124006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6124006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6124006 Supplier Thermo Scientific Chemicals Supplier No. H6124006

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73821-95-1
Molecular Formula C15H25NO6
Molecular Weight (g/mol) 315.366
MDL Number MFCD00061996
InChI Key NLPQIWFEEKQBBN-NSHDSACASA-N
Synonym boc-asp ochx-oh, boc-asp ochex-oh, boc-l-aspartic acid 4-cyslohexyl ester, n-boc-l-aspartic acid 4-cyclohexyl ester, 2s-2-tert-butoxycarbonyl amino-4-cyclohexyloxy-4-oxobutanoic acid, boc-asp o-chex, ambotzbaa1074, pubchem12097, 4-cyclohexyl n-boc-l-aspartate, boc-l-aspartic acid b-cyclohexyl ester
PubChem CID 7009416
IUPAC Name (2S)-4-cyclohexyloxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)OC1CCCCC1)C(=O)O

Specifications

Melting Point 93°C to 95°C
Color White
Beilstein 3563575
Quantity 5 g
Formula Weight 315.36
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Boc-L-aspartic acid 4-cyclohexyl ester
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.