Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-L-aspartic acid, 98%

Catalog No. p-7044856
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0849803 1 g
AAL0849806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0849803 Supplier Thermo Scientific Chemicals Supplier No. L0849803
Only null left
Add to Cart

CAS: 13726-67-5 | C9H15NO6 | 233.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13726-67-5
Molecular Formula C9H15NO6
Molecular Weight (g/mol) 233.22
MDL Number MFCD00037279
InChI Key KAJBMCZQVSQJDE-YFKPBYRVSA-N
Synonym boc-asp-oh, n-tert-butoxycarbonyl-l-aspartic acid, boc-l-aspartic acid, boc-l-asp-oh, n-boc-l-aspartic acid, boc-aspartic acid, boc-l-aspartic acid-oh, n-tert-butyloxycarbonyl-l-aspartic acid, n-t-butoxycarbonyl-l-aspatic acid, 2s-2-tert-butoxycarbonyl amino butanedioic acid
PubChem CID 99718
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanedioic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)O

Specifications

Melting Point 115°C
Quantity 1 g
Beilstein 1913973
Optical Rotation −5.5° (c=1 in Methanol)
Formula Weight 233.23
Percent Purity 98%
Chemical Name or Material N-Boc-L-aspartic acid
EINECSNumber 237-294-7
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.