Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-L-glutamic acid 1-tert-butyl ester, 99%

Catalog No. AAH5984203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5984203 1 g
AAH5984206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5984203 Supplier Thermo Scientific Chemicals Supplier No. H5984203
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 24277-39-2 | C14H25NO6 | 303.36 g/mol

Boc-Glu-OtBu is an N-terminal protected amino acid used in solid-phase peptide synthesis (SPPS) to make unique peptides containing glutamate tert-butyl ester residues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Boc-Glu-OtBu is an N-terminal protected amino acid used in solid-phase peptide synthesis (SPPS) to make unique peptides containing glutamate tert-butyl ester residues.

Solubility
Soluble in dimethyl formamide.

Notes
Recommended storage temperature: -20°C. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24277-39-2
Molecular Formula C14H25NO6
Molecular Weight (g/mol) 303.36
MDL Number MFCD00038273
InChI Key YMOYURYWGUWMFM-UHFFFAOYNA-N
Synonym boc-glu-otbu, boc-l-glutamic acid 1-tert-butyl ester, s-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, n-t-boc-l-glutamic acid a-t-butyl-ester, s-5-tert-butoxy-4-tert-butoxycarbonylamino-5-oxopentanoic acid, 4s-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, t-boc-glu-otbu, n-boc-l-glutamic acid 1-tert-butyl ester, pubchem19036, ksc495g0n
PubChem CID 6993444
SMILES CC(C)(C)OC(=O)NC(CCC(O)=O)C(=O)OC(C)(C)C

Specifications

Quantity 1 g
Beilstein 3653769
Solubility Information Soluble in dimethyl formamide.
IUPAC Name 5-(tert-butoxy)-4-{[(tert-butoxy)carbonyl]amino}-5-oxopentanoic acid
Formula Weight 303.36
Percent Purity 99%
Chemical Name or Material N-Boc-L-glutamic acid 1-tert-butyl ester
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.