Learn More
Medchemexpress LLC N-Boc-L-Prolinal | 69610-41-9 | 98.8% | C10H17NO3 | 25 G
SDP
Catalog No. 5000358929
Encompass_Preferred
N-Boc-L-Prolinal is a proline derivative featuring a Boc protecting group, commonly employed in the synthesis of biologically active peptide mimetics. This compound is primarily intended for research applications, including the development of Dolastatin 10 analogs that exhibit anti-colon cancer activity.
  • Proline derivative with a Boc protecting group
  • Used in the synthesis of biologically active peptide mimetics
  • Applied in the development of Dolastatin 10 analogs
  • Analogs demonstrate anti-colon cancer activity
  • Intended for research use only
Catalog No. 50-003-58929 Supplier Medchemexpress LLC Supplier No. 25GHY78927
N-Boc-L-Prolinal is a proline derivative featuring a Boc protecting group, commonly employed in the synthesis of biologically active peptide mimetics. This compound is primarily intended for research applications, including the development of Dolastatin 10 analogs that exhibit anti-colon cancer activity.
  • Proline derivative with a Boc protecting group
  • Used in the synthesis of biologically active peptide mimetics
  • Applied in the development of Dolastatin 10 analogs
  • Analogs demonstrate anti-colon cancer activity
  • Intended for research use only