Learn More
Medchemexpress LLC N-Boc-L-pyroglutamic acid | 53100-44-0 | 99.7% | 229.23 | 1 G
SDP
Catalog No. 5000366914
Encompass_Preferred
Boc-Pyr-OH is a Boc-protected pyroglutamic acid derivative. It stabilizes the amino group of pyroglutamic acid through the Boc protecting group, preventing unwanted reactions during peptide synthesis. It can be used in the research of thyrotropin-releasing hormone (TRH) synthesis.
  • Boc-protected pyroglutamic acid derivative
  • Stabilizes amino group
  • Prevents unwanted reactions during peptide synthesis
  • Used in research of thyrotropin-releasing hormone (TRH) synthesis
Catalog No. 50-003-66914 Supplier Medchemexpress LLC Supplier No. HYW0096731G
Boc-Pyr-OH is a Boc-protected pyroglutamic acid derivative. It stabilizes the amino group of pyroglutamic acid through the Boc protecting group, preventing unwanted reactions during peptide synthesis. It can be used in the research of thyrotropin-releasing hormone (TRH) synthesis.
  • Boc-protected pyroglutamic acid derivative
  • Stabilizes amino group
  • Prevents unwanted reactions during peptide synthesis
  • Used in research of thyrotropin-releasing hormone (TRH) synthesis