Learn More
Medchemexpress LLC N-Boc-L-pyroglutamic acid | 53100-44-0 | 99.7% | 229.23 | 5 G
SDP
Catalog No. 5000453130
Encompass_Preferred
Boc-Pyr-OH is a Boc-protected pyroglutamic acid derivative. It stabilizes the amino group of pyroglutamic acid through the Boc protecting group, which prevents unwanted reactions during peptide synthesis. This compound can be used in the research of thyrotropin-releasing hormone (TRH) synthesis. It is for research use only.
  • Boc-protected pyroglutamic acid derivative
  • Stabilizes the amino group of pyroglutamic acid
  • Prevents unwanted reactions during peptide synthesis
  • Applicable in thyrotropin-releasing hormone (TRH) synthesis research
Catalog No. 50-004-53130 Supplier Medchemexpress LLC Supplier No. HYW0096735G
Boc-Pyr-OH is a Boc-protected pyroglutamic acid derivative. It stabilizes the amino group of pyroglutamic acid through the Boc protecting group, which prevents unwanted reactions during peptide synthesis. This compound can be used in the research of thyrotropin-releasing hormone (TRH) synthesis. It is for research use only.
  • Boc-protected pyroglutamic acid derivative
  • Stabilizes the amino group of pyroglutamic acid
  • Prevents unwanted reactions during peptide synthesis
  • Applicable in thyrotropin-releasing hormone (TRH) synthesis research