Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-L-tryptophanol, 95%, Thermo Scientific Chemicals

Catalog No. AAH6657103
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6657103 1 g
AAH66571MD 250 mg
AAH6657106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6657103 Supplier Thermo Scientific Chemicals Supplier No. H6657103

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82689-19-8
Molecular Formula C16H22N2O3
Molecular Weight (g/mol) 290.36
MDL Number MFCD00235953
InChI Key JEFQUFUAEKORKL-LBPRGKRZSA-N
Synonym n-boc-l-tryptophanol, boc-tryptophanol, boc-l-tryptophanol, boc-trp-ol, s-tert-butyl 1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate, s-3-2-boc-amino-3-hydroxypropyl-indole, boc-d-trp-ol, n-alpha-boc-l-tryptophanol, boc-trypotophanol, boc-l-trp-ol
PubChem CID 7019533
IUPAC Name tert-butyl N-[(2S)-1-hydroxy-3-(1H-indol-3-yl)propan-2-yl]carbamate
SMILES CC(C)(C)OC(=O)N[C@H](CO)CC1=CNC2=CC=CC=C12

Specifications

Melting Point 118°C to 122°C
Quantity 1 g
Formula Weight 290.36
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-L-tryptophanol
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.