Learn More
Medchemexpress LLC N-Boc-N-bis(PEG2-acid) | 2054339-01-2 | 99.9% | C19H35NO10 | 100 MG
SDP
Catalog No. 5000356225
Encompass_Preferred
N-Boc-N-bis(PEG2-acid) is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs are compounds that contain two different ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other targeting a protein of interest. This mechanism allows PROTACs to exploit the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • Functions as a PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates targeted protein degradation
  • For research use only
Catalog No. 50-003-56225 Supplier Medchemexpress LLC Supplier No. HY140538100MG
N-Boc-N-bis(PEG2-acid) is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs are compounds that contain two different ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other targeting a protein of interest. This mechanism allows PROTACs to exploit the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • Functions as a PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates targeted protein degradation
  • For research use only