Learn More
Medchemexpress LLC N-Boc-N-bis(PEG2-acid) | 2054339-01-2 | 99.9% | C19H35NO10 | 250 MG
SDP
Catalog No. 5000352717
Encompass_Preferred
N-Boc-N-bis(PEG2-acid) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. This product is for research use only and not sold to patients. PROTACs are compounds that utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • For research use only
  • Utilizes the intracellular ubiquitin-proteasome system to selectively degrade target proteins
  • Store in pure form at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-003-52717 Supplier Medchemexpress LLC Supplier No. 250MGHY140538
N-Boc-N-bis(PEG2-acid) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. This product is for research use only and not sold to patients. PROTACs are compounds that utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • For research use only
  • Utilizes the intracellular ubiquitin-proteasome system to selectively degrade target proteins
  • Store in pure form at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month