Learn More
Medchemexpress LLC N-Boc-N-bis(PEG2-acid) | 2054339-01-2 | 99.9% | C19H35NO10 | 500 MG
SDP
Catalog No. 5000352944
Encompass_Preferred
N-Boc-N-bis(PEG2-acid) is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. These molecules exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins, making them suitable for research applications.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-52944 Supplier Medchemexpress LLC Supplier No. HY140538500MG
N-Boc-N-bis(PEG2-acid) is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. These molecules exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins, making them suitable for research applications.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only