Learn More
Medchemexpress LLC N-Boc-N-bis(PEG2-propargyl) | 2100306-86-1 | 98.0% | 369.45 | 25 MG
SDP
Catalog No. 5000351697
Encompass_Preferred
N-Boc-N-bis(PEG2-propargyl) is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an alkyne group capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Capable of copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups
  • For research use only
Catalog No. 50-003-51697 Supplier Medchemexpress LLC Supplier No. 25MGHY140089
N-Boc-N-bis(PEG2-propargyl) is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an alkyne group capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Capable of copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups
  • For research use only