Learn More
Medchemexpress LLC N-boc-n-bis(PEG4-OH) | 2093154-01-7 | 469.57 g/mol | C21H43NO10 | 25 MG
SDP
Catalog No. 5000202146
Encompass_Preferred
N-boc-n-bis(PEG4-OH) is a PEG-based cleavable linker used in the synthesis of proteolysis-targeting chimeras (PROTACs) and antibody-drug conjugates (ADCs). It provides a Boc-protected amine for functionalization and bis(PEG4) arms for flexibility and aqueous solubility.
  • Boc-protected amine provides a synthetic handle for derivatization.
  • Bis(PEG4) arms increase flexibility and solubility.
  • Suitable as a cleavable linker in ADC construction.
  • Commonly used as a spacer in PROTAC synthesis.
  • Supplied in multiple package sizes to suit research needs.
Catalog No. 50-002-02146 Supplier Medchemexpress LLC Supplier No. HY13044925MG
N-boc-n-bis(PEG4-OH) is a PEG-based cleavable linker used in the synthesis of proteolysis-targeting chimeras (PROTACs) and antibody-drug conjugates (ADCs). It provides a Boc-protected amine for functionalization and bis(PEG4) arms for flexibility and aqueous solubility.
  • Boc-protected amine provides a synthetic handle for derivatization.
  • Bis(PEG4) arms increase flexibility and solubility.
  • Suitable as a cleavable linker in ADC construction.
  • Commonly used as a spacer in PROTAC synthesis.
  • Supplied in multiple package sizes to suit research needs.