Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-N-methyl-L-phenylglycine, 97%, Thermo Scientific™

Catalog No. AAH63720MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63720MD 250 mg
AAH6372003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH63720MD Supplier Thermo Scientific Chemicals Supplier No. H63720MD

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30925-11-2
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.31
MDL Number MFCD00038799
InChI Key COABPHLHHQAKPL-UHFFFAOYNA-N
Synonym boc-n-me-phg-oh, boc-n-methyl-l-phenylglycine, s-tert-butoxycarbonyl methyl amino phenyl acetic acid, s-2-tert-butoxycarbonyl methyl amino-2-phenylacetic acid, ambotzbaa1259, pubchem12248, boc-n-methyl-s-2-phenylglycine, s-methyl tert-butoxycarbonyl amino phenylacetic acid, n-alpha-t-butyloxycarbonyl-n-alpha-methyl-l-phenylglycine, 2s-tert-butoxycarbonyl methyl amino phenyl acetic acid
PubChem CID 7010620
SMILES CN(C(C(O)=O)C1=CC=CC=C1)C(=O)OC(C)(C)C

Specifications

IUPAC Name (2S)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]-2-phenylacetic acid
Quantity 250 mg
Formula Weight 265.3
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-Boc-N-methyl-L-phenylglycine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.