Learn More
Medchemexpress LLC N-Boc-O-tosyl hydroxylamine | 105838-14-0 | 287.33 | 1 G
SDP
Catalog No. 5000446317
Encompass_Preferred
N-Boc-O-tosyl hydroxylamine serves as a safe and efficient nitrogen source for the N-amination of aryl and alkyl amines. It promotes electrophilic N-amination of aryl amines and alkyl amines to generate β-Boc-protected aryl hydrazines and alkyl hydrazines. This product is intended for research use only.
  • Functions as a safe and efficient nitrogen source
  • Promotes electrophilic N-amination
  • Generates β-Boc-protected aryl hydrazines and alkyl hydrazines
  • Provided as a solid
  • Color is white to off-white
Catalog No. 50-004-46317 Supplier Medchemexpress LLC Supplier No. HYW0004381G
N-Boc-O-tosyl hydroxylamine serves as a safe and efficient nitrogen source for the N-amination of aryl and alkyl amines. It promotes electrophilic N-amination of aryl amines and alkyl amines to generate β-Boc-protected aryl hydrazines and alkyl hydrazines. This product is intended for research use only.
  • Functions as a safe and efficient nitrogen source
  • Promotes electrophilic N-amination
  • Generates β-Boc-protected aryl hydrazines and alkyl hydrazines
  • Provided as a solid
  • Color is white to off-white