Learn More
Medchemexpress LLC N-Boc-O-tosyl hydroxylamine | 105838-14-0 | 287.33 | 25 G
SDP
Catalog No. 5000449949
Encompass_Preferred
N-Boc-O-tosyl hydroxylamine is a safe and efficient nitrogen source for N-amination of aryl and alkyl amines. It promotes electrophilic N-amination of aryl and alkyl amines to generate β-Boc-protected aryl hydrazines and alkyl hydrazines.
  • Safe nitrogen source
  • Efficient for N-amination of aryl and alkyl amines
  • Promotes electrophilic N-amination
  • Generates β-Boc-protected aryl hydrazines
  • Generates β-Boc-protected alkyl hydrazines
Catalog No. 50-004-49949 Supplier Medchemexpress LLC Supplier No. 25GHYW000438
N-Boc-O-tosyl hydroxylamine is a safe and efficient nitrogen source for N-amination of aryl and alkyl amines. It promotes electrophilic N-amination of aryl and alkyl amines to generate β-Boc-protected aryl hydrazines and alkyl hydrazines.
  • Safe nitrogen source
  • Efficient for N-amination of aryl and alkyl amines
  • Promotes electrophilic N-amination
  • Generates β-Boc-protected aryl hydrazines
  • Generates β-Boc-protected alkyl hydrazines