Learn More
Medchemexpress LLC N-Boc-O-tosyl hydroxylamine | 105838-14-0 | 287.33 | 500 MG
Supplier: Medchemexpress LLC HYW000438500MG
N-Boc-O-tosyl hydroxylamine is a safe and efficient nitrogen source for N-amination of aryl and alkyl amines. It promotes electrophilic N-amination of aryl amines and alkyl amines to generate β-Boc-protected aryl hydrazines and alkyl hydrazines.
- Safe and efficient nitrogen source
- Promotes electrophilic N-amination of aryl and alkyl amines
- Generates β-Boc-protected aryl and alkyl hydrazines