Learn More
Medchemexpress LLC N-Boc-O-tosyl hydroxylamine | 105838-14-0 | MFCD22201097 | 287.33 | 5 G
SDP
Catalog No. 5000447649
Encompass_Preferred
N-Boc-O-tosyl hydroxylamine is a safe and efficient nitrogen source for N-amination of aryl and alkyl amines. It promotes electrophilic N-amination of aryl amines and alkyl amines to generate β-Boc-protected aryl hydrazines and alkyl hydrazines.
  • Safe and efficient nitrogen source
  • Promotes electrophilic N-amination of aryl amines
  • Promotes electrophilic N-amination of alkyl amines
  • Generates β-Boc-protected aryl hydrazines
  • Generates β-Boc-protected alkyl hydrazines
Catalog No. 50-004-47649 Supplier Medchemexpress LLC Supplier No. HYW0004385G
N-Boc-O-tosyl hydroxylamine is a safe and efficient nitrogen source for N-amination of aryl and alkyl amines. It promotes electrophilic N-amination of aryl amines and alkyl amines to generate β-Boc-protected aryl hydrazines and alkyl hydrazines.
  • Safe and efficient nitrogen source
  • Promotes electrophilic N-amination of aryl amines
  • Promotes electrophilic N-amination of alkyl amines
  • Generates β-Boc-protected aryl hydrazines
  • Generates β-Boc-protected alkyl hydrazines