Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Boc-P-Phenylenediamine

Catalog No. 154855G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-485-5G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 15-485-5G Supplier Sigma Aldrich Supplier No. 154855G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-Boc-p-phenylenediamine can be used as:A reactant to prepare perylene monoimide-based dyes for dye-sensitized solar cell applications.A starting material to synthesize covalent organic frameworks, which are used as proton exchange membranes for hydrogen fuel cell applications.A reactant in the synthesis of bestatin derived hydroxamic acids as potent pan-HDAC inhibitors.Mono-N-acylated diamine, precursor of different Para-substituted anilines (e.g., 4-azido aniline); Synthesis of photoactivable derivative of ouabaine.

Specifications

CAS 402846-78-0
Melting Point 37°C
Density 1.198 g/mL at 20°C
Molecular Formula C9H17BF4N2
Linear Formula C9H17BF4N2
MDL Number MFCD03427618
Quantity 50 g
Synonym 4-(tert-Butoxycarbonylamino)aniline; tert-Butyl-4-aminophenylcarbamate
Molecular Weight (g/mol) 240.05 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.