Learn More
Medchemexpress LLC N-Boc-PEG1-bromide | 164332-88-1 | 97.9% | 268.15 | C9H18BrNO3 | 250 MG
SDP
Catalog No. 5000203912
Encompass_Preferred
N-Boc-PEG1-bromide is a Boc-protected PEG1 bromide reagent used as a short, hydrophilic linker for conjugation chemistry, including antibody-drug conjugate (ADC) and PROTAC linker synthesis. It provides a terminal bromide for alkylation or nucleophilic substitution and a Boc-protected amino group for orthogonal protection strategies.
  • Provides a short PEG spacer to increase solubility.
  • Contains a terminal bromide for alkylation and substitution reactions.
  • Includes a Boc-protected amino group for orthogonal deprotection.
  • Used in ADC and PROTAC linker synthesis and general conjugation chemistry.
  • Supplied in multiple package sizes for research use.
Catalog No. 50-002-03912 Supplier Medchemexpress LLC Supplier No. HY130503250MG
N-Boc-PEG1-bromide is a Boc-protected PEG1 bromide reagent used as a short, hydrophilic linker for conjugation chemistry, including antibody-drug conjugate (ADC) and PROTAC linker synthesis. It provides a terminal bromide for alkylation or nucleophilic substitution and a Boc-protected amino group for orthogonal protection strategies.
  • Provides a short PEG spacer to increase solubility.
  • Contains a terminal bromide for alkylation and substitution reactions.
  • Includes a Boc-protected amino group for orthogonal deprotection.
  • Used in ADC and PROTAC linker synthesis and general conjugation chemistry.
  • Supplied in multiple package sizes for research use.