Learn More
Medchemexpress LLC N-(Boc-PEG1)-N-bis(PEG2-propargyl) | 2100306-63-4 | 456.57 | 25 MG
SDP
Catalog No. 5000351408
Encompass_Preferred
N-(Boc-PEG1)-N-bis(PEG2-propargyl) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Click chemistry reagent with an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Facilitates selective degradation of target proteins
Catalog No. 50-003-51408 Supplier Medchemexpress LLC Supplier No. 25MGHY140090
N-(Boc-PEG1)-N-bis(PEG2-propargyl) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Click chemistry reagent with an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Facilitates selective degradation of target proteins