Learn More
Medchemexpress LLC N-(Boc-PEG1)-N-bis(PEG2-propargyl) | 2100306-63-4 | 456.57 | 250 MG
SDP
Catalog No. 5000348859
Encompass_Preferred
N-(Boc-PEG1)-N-bis(PEG2-propargyl) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It also functions as a click chemistry reagent, featuring an alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
Catalog No. 50-003-48859 Supplier Medchemexpress LLC Supplier No. 250MGHY140090
N-(Boc-PEG1)-N-bis(PEG2-propargyl) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It also functions as a click chemistry reagent, featuring an alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)