Learn More
Medchemexpress LLC N-(Boc-PEG1)-N-bis(PEG2-propargyl) | 2100306-63-4 | 95-98% | 456.57 | 100 MG
SDP
Catalog No. 5000350511
Encompass_Preferred
N-(Boc-PEG1)-N-bis(PEG2-propargyl) is a PEG-based PROTAC linker used in the synthesis of PROTACs. It also functions as a click chemistry reagent, featuring an Alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Azide-containing molecules.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group
  • Capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc)
Catalog No. 50-003-50511 Supplier Medchemexpress LLC Supplier No. HY140090100MG
N-(Boc-PEG1)-N-bis(PEG2-propargyl) is a PEG-based PROTAC linker used in the synthesis of PROTACs. It also functions as a click chemistry reagent, featuring an Alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Azide-containing molecules.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group
  • Capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc)