Learn More
Medchemexpress LLC N-Boc-PEG10-alcohol | 2238844-74-9 | MFCD08457831 | 98.0% | 557.67 g/mol | C25H51NO12 | 250 MG
SDP
Catalog No. 5000283240
Encompass_Preferred
N-Boc-PEG10-alcohol is a Boc-protected polyethylene glycol (PEG10) alcohol used as a linker in proteolysis targeting chimera (PROTAC) and antibody-drug conjugate (ADC) synthesis. It supplies a hydrophilic spacer with a protected terminal amine that can be selectively deprotected and derivatized for conjugation. Supplied for research use; request the certificate of analysis and safety data for QC and handling details.
  • Provides a hydrophilic PEG10 spacer for improved solubility.
  • Boc-protected terminal amine enables selective deprotection and coupling.
  • Suitable for use in PROTAC and ADC linker design and medicinal chemistry.
  • Available in multiple small-scale quantities for research applications.
  • QC report and safety data available upon request.
Catalog No. 50-002-83240 Supplier Medchemexpress LLC Supplier No. HYW190857250MG
N-Boc-PEG10-alcohol is a Boc-protected polyethylene glycol (PEG10) alcohol used as a linker in proteolysis targeting chimera (PROTAC) and antibody-drug conjugate (ADC) synthesis. It supplies a hydrophilic spacer with a protected terminal amine that can be selectively deprotected and derivatized for conjugation. Supplied for research use; request the certificate of analysis and safety data for QC and handling details.
  • Provides a hydrophilic PEG10 spacer for improved solubility.
  • Boc-protected terminal amine enables selective deprotection and coupling.
  • Suitable for use in PROTAC and ADC linker design and medicinal chemistry.
  • Available in multiple small-scale quantities for research applications.
  • QC report and safety data available upon request.