Learn More
Medchemexpress LLC N-(Boc-PEG3)-N-bis(PEG2-alcohol) | 2055042-60-7 | > 96.0% | C25H52N2O11 | 100 MG
SDP
Catalog No. 5000355825
Encompass_Preferred
N-(Boc-PEG3)-N-bis(PEG2-alcohol) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These PROTACs consist of two distinct ligands connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets a specific protein.
  • Composed of two distinct ligands connected by a linker.
  • One ligand targets an E3 ubiquitin ligase and the other targets a specific protein.
  • Leverages the intracellular ubiquitin-proteasome system.
  • Achieves selective degradation of target proteins.
Catalog No. 50-003-55825 Supplier Medchemexpress LLC Supplier No. HY141254100MG
N-(Boc-PEG3)-N-bis(PEG2-alcohol) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These PROTACs consist of two distinct ligands connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets a specific protein.
  • Composed of two distinct ligands connected by a linker.
  • One ligand targets an E3 ubiquitin ligase and the other targets a specific protein.
  • Leverages the intracellular ubiquitin-proteasome system.
  • Achieves selective degradation of target proteins.