Learn More
Medchemexpress LLC N-(Boc-PEG3)-N-bis(PEG2-alcohol) | 2055042-60-7 | C25H52N2O11 | 250 MG
SDP
Catalog No. 5000352898
Encompass_Preferred
N-(Boc-PEG3)-N-bis(PEG2-alcohol) is a PEG-based PROTAC linker used in the synthesis of Proteolysis Targeting Chimeras (PROTACs). PROTACs are designed to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Enables targeted protein degradation via the ubiquitin-proteasome system
  • Facilitates connection between an E3 ubiquitin ligase ligand and a target protein ligand
Catalog No. 50-003-52898 Supplier Medchemexpress LLC Supplier No. 250MGHY141254
N-(Boc-PEG3)-N-bis(PEG2-alcohol) is a PEG-based PROTAC linker used in the synthesis of Proteolysis Targeting Chimeras (PROTACs). PROTACs are designed to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Enables targeted protein degradation via the ubiquitin-proteasome system
  • Facilitates connection between an E3 ubiquitin ligase ligand and a target protein ligand