Learn More
Medchemexpress LLC N-(Boc-PEG3)-N-bis(PEG3-acid) | 2055042-61-8 | 97.0% | C31H60N2O15 | 25 MG
SDP
Catalog No. 5000357391
Encompass_Preferred
N-(Boc-PEG3)-N-bis(PEG3-acid) is a PEG-based PROTAC linker. It is designed for use in the synthesis of PROTACs. This product is for research use only.
  • PEG-based PROTAC linker
  • For use in the synthesis of PROTACs
  • For research use only
Catalog No. 50-003-57391 Supplier Medchemexpress LLC Supplier No. 25MGHY140536
N-(Boc-PEG3)-N-bis(PEG3-acid) is a PEG-based PROTAC linker. It is designed for use in the synthesis of PROTACs. This product is for research use only.
  • PEG-based PROTAC linker
  • For use in the synthesis of PROTACs
  • For research use only