Learn More
Medchemexpress LLC N-Boc-PEG4-bromide | 1076199-21-7 | MFCD09840081 | 98.0% | 356.25 g/mol | C13H26BrNO5 | 100 MG
SDP
Catalog No. 5000202925
Encompass_Preferred
N-Boc-PEG4-bromide is a Boc-protected PEG4 linker bearing a terminal bromide. It is supplied for research use as a cleavable linker building block for antibody-drug conjugate (ADC) synthesis and as a PEG-based spacer in proteolysis-targeting chimera (PROTAC) assembly; the terminal bromide enables alkylation chemistry while the PEG spacer improves solubility and flexibility.
  • Boc-protected amino group for protecting amines during synthesis.
  • Terminal bromide enables nucleophilic substitution and alkylation.
  • Peg4 spacer increases aqueous solubility and provides flexibility.
  • High purity for research applications.
  • Available in small pack sizes suitable for lab-scale synthesis.
Catalog No. 50-002-02925 Supplier Medchemexpress LLC Supplier No. HYW046471100MG
N-Boc-PEG4-bromide is a Boc-protected PEG4 linker bearing a terminal bromide. It is supplied for research use as a cleavable linker building block for antibody-drug conjugate (ADC) synthesis and as a PEG-based spacer in proteolysis-targeting chimera (PROTAC) assembly; the terminal bromide enables alkylation chemistry while the PEG spacer improves solubility and flexibility.
  • Boc-protected amino group for protecting amines during synthesis.
  • Terminal bromide enables nucleophilic substitution and alkylation.
  • Peg4 spacer increases aqueous solubility and provides flexibility.
  • High purity for research applications.
  • Available in small pack sizes suitable for lab-scale synthesis.